2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16092-14
A16092-22
A16092-36
CAAAA16092-14EA
66.83
CAD
CAAAA16092-14
CAAAA16092-22
CAAAA16092-36
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid 99%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
MDL: MFCD00007533
Beilstein: 1781572
Soluble in water
Purity: 99%
CAS No.: 10191-18-1
Molecular Formula: C6H15NO5S
Formula Weight: 213.25
Melting Point: 153-157°
Solubility: Soluble in water
Beilstein Reference: 1781572
MDL No.: MFCD00007533
Formula:
C₆H₁₅NO₅S Boiling Pt: 405 °C (1013 hPa) Melting Pt: 153…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007533 CAS Number: 10191-18-1 EINECS: 233-465-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...