Tributyl phosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TBP, TBPA

A16084-0J A16084-AK A16084-AU
CAAAA16084-0JEA 385.52 CAD
CAAAA16084-0J CAAAA16084-AK CAAAA16084-AU
Tributyl phosphate 98%
Tributyl phosphate

Purity: 98% CAS No.: 126-73-8 Molecular Formula: C12H27O4P Formula Weight: 266.32 Flash Point: 152°(305°F) Physical Form: Liquid Density: 0.977 Refractive Index: 1.4240 Appearance: Colorless Odor: Odorless Melting Point: -80° Boiling Point: 155-156°/10mm Beilstein Reference: 1710584 Merck Reference: 14,9618 MDL No.: MFCD00009436

Formula: (CH₃(CH₂)₃O)₃PO
MW: 266.32 g/mol
Boiling Pt: 155…156 °C/10 mm
Melting Pt: –80 °C
Density: 0.977
Flash Pt: 152 °C(305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 14,9618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR