Diethylene glycol monobutyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

A16068-0J A16068-AU
CAAAA16068-0JEA 199.08 CAD
CAAAA16068-0J CAAAA16068-AU
Diethylene glycol monobutyl ether 99%
Diethylene glycol monobutyl ether

Purity: 99% CAS No.: 112-34-5 Molecular Formula: C8H18O3 Formula Weight: 162.23 Flash Point: 100°(212°F) Refractive Index: 1.4320 Odor: schwach, mild Melting Point: -68° Boiling Point: 230-232° Merck Reference: 14,1557 MDL No.: MFCD00002881

Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4
Melting Pt: -68
Density: 0.967
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 14,1557

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR