Diethylene glycol monomethyl ether 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether
Stabilization:
50-150 mg/kg Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A16063-0F
A16063-AP
CAAAA16063-0FEA
138.09
CAD
CAAAA16063-0F
CAAAA16063-AP
Diethylene glycol monomethyl ether 98% stabilised
Diethylene glycol monomethyl ether
Purity: 98%
CAS No.: 111-77-3
Molecular Formula: C5H12O3
Formula Weight: 120.15
Flash Point: 83°(181°F)
Refractive Index: 1.4260
Odor: Light
Melting Point: -70°
Boiling Point: 194°
Beilstein Reference: 1697812
Merck Reference: 14,6037
MDL No.: MFCD00002871
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol Boiling Pt: 193 Melting Pt: -65 Density: 1.010 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002871 CAS Number: 111-77-3 EINECS: 203-906-6 Merck Index: 14,6037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...