Methylcyclohexane 99%

Supplier: Thermo Scientific Chemicals
Danger

A16057-0F A16057-AP
CAAAAA16057-AEEA 67.4 CAD
CAAAAA16057-AE CAAAA16057-0F CAAAA16057-AP
Methylcyclohexane 99%
Methylcyclohexane

Purity: 99% CAS No.: 108-87-2 Molecular Formula: C7H14 Formula Weight: 98.19 Flash Point: -4°(-25°F) Refractive Index: 1.4230 Melting Point: -126° Boiling Point: 101° Beilstein Reference: 505972 Merck Reference: 14,6047 UN No.: 2296 MDL No.: MFCD00001497

Formula: C₆H₁₁CH₃
MW: 98.19 g/mol
Boiling Pt: 101 °C
Melting Pt: –126 °C
Density: 0.771
Flash Pt: –4 °C (–25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 14,6047
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR