Indigo carmine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CI 73015, 5,5'-Indigodisulfonic acid, Disodium salt, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulfonate, Carmine Blue

A16052-14 A16052-22
CAAAA16052-14EA 67.78 CAD
CAAAA16052-14 CAAAA16052-22
Indigo carmine
Indigo carmine

Useful in the staining of Negri bodies


CAS No.: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Formula Weight: 466.36 Odor: Odorless Melting Point: 300° Beilstein Reference: 4103904 Merck Reference: 14,4944 MDL No.: MFCD00005723

Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
EINECS: 212-728-8
Merck Index: 14,4944

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR