4-Phenoxyaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16048-14
A16048-22
A16048-36
CAAAA16048-14EA
49.14
CAD
CAAAA16048-14
CAAAA16048-22
CAAAA16048-36
4-Phenoxyaniline 97%
4-Phenoxyaniline
Purity: 97%
CAS No.: 139-59-3
Molecular Formula: C12H11NO
Formula Weight: 185.23
Melting Point: 82-86°
Boiling Point: 140°/2mm
UN No.: 3077
MDL No.: MFCD00007862
Formula:
C₁₂H₁₁NO Boiling Pt: 140 °C/2 mm Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007862 CAS Number: 139-59-3 EINECS: 205-367-2 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...