TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16046-0F
A16046-AE
A16046-AP
CAAAA16046-0FEA
293.12
CAD
CAAAA16046-0F
CAAAA16046-AE
CAAAA16046-AP
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Detergent, equivalent to Nonidet P-40
CAS No.: 9002-93-1
Formula Weight: ca. 625
Flash Point: 180°(356°F)
Physical Form: Liquid
Density: 1.061
Refractive Index: 1.4900
Odor: Mild
Melting Point: ca 5°
Boiling Point:
Detergent, equivalent to Nonidet P-40
Boiling Pt:
>250 °C Melting Pt: ∼ 5 °C Density: 1.061 Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00132505 CAS Number: 9002-93-1 UN: 3082 ADR: 9,III Merck Index: 14,6761 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...