Indole-3-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A16041-03 A16041-06
CAAAAA16041-14EA 843.59 CAD
CAAAAA16041-14 CAAAA16041-03 CAAAA16041-06
Indole-3-carbonitrile 98%
Indole-3-carbonitrile

Purity: 98% CAS No.: 5457-28-3 Molecular Formula: C9H6N2 Formula Weight: 142.16 Odor: Odorless Melting Point: 177-182° Beilstein Reference: 120888 MDL No.: MFCD00022717

Formula: C₉H₆N₂
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00022717
CAS Number: 5457-28-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR