4-Fluoro-2-methylphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16032-06
A16032-14
CAAAAA16032-22EA
2802.05
CAD
CAAAAA16032-22
CAAAA16032-06
CAAAA16032-14
4-Fluoro-2-methylphenol 98%
4-Fluoro-2-methylphenol
Purity: 98%
CAS No.: 452-72-2
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 85°(185°F)
Refractive Index: 1.5165
Odor: Phenol-like
Melting Point: 32-36°
Boiling Point: 87°/14mm
Beilstein Reference: 2041490
UN No.: 3261
MDL No.: MFCD00075088
Formula:
C₇H₇FO Boiling Pt: 87 °C/14 mm Melting Pt: 32…36 °C Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075088 CAS Number: 452-72-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...