4'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoro-2-hydroxyacetophenone
A16030-06
A16030-14
CAAAA16030-06EA
101.91
CAD
CAAAA16030-06
CAAAA16030-14
4'-Fluoro-2'-hydroxyacetophenone 98%
4'-Fluoro-2'-hydroxyacetophenone
Purity: 98%
CAS No.: 1481-27-2
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point: 98°(208°F)
Melting Point: 29-34°
Boiling Point: 65°/2mm
Beilstein Reference: 2249799
MDL No.: MFCD00143203
Formula:
C₈H₇FO₂ MW: 154.14 Boiling Pt: 65 °C (2 mmHg) Melting Pt: 29…34 °C Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00143203 CAS Number: 1481-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...