4-Methoxythiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Methoxythiophenol, 4-Mercaptoanisol, 1-Methoxy-4-mercaptobenzene, 4-Methoxybenzenethiol, 4-Methoxybenzene-1-thiol, 4-Methoxyphenyl mercaptan

A16009-06 A16009-14
CAAAAA16009-22EA 1347.6 CAD
CAAAAA16009-22 CAAAA16009-06 CAAAA16009-14
4-Methoxythiophenol 97%
4-Methoxythiophenol

Purity: 97% CAS No.: 696-63-9 Molecular Formula: C7H8OS Formula Weight: 140.21 Flash Point: 96°(204°F) Refractive Index: 1.5831 Odor: Unpleasant Boiling Point: 100-103°/13mm Beilstein Reference: 1446910 MDL No.: MFCD00004849

Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 228
Melting Pt: 42
Density: 1.140
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR