1,3-Dichloro-2-propanol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16003-0B
A16003-30
CAAAA16003-0BEA
202.24
CAD
CAAAA16003-0B
CAAAA16003-30
1,3-Dichloro-2-propanol 98+%
1,3-Dichloro-2-propanol
Purity: 98+%
CAS No.: 96-23-1
Molecular Formula: C3H6Cl2O
Formula Weight: 128.99
Flash Point: 85°(185°F)
Refractive Index: 1.4830
Melting Point: -4°
Boiling Point: 174-176°
Beilstein Reference: 1732063
Merck Reference: 14,3074
UN No.: 2750
MDL No.: MFCD00000951
Formula:
ClH₂CCHOHCH₂Cl Boiling Pt: 174…176 °C Melting Pt: –4 °C Density: 1.366 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000951 CAS Number: 96-23-1 EINECS: 202-491-9 UN: 2750 ADR: 6.1,II Merck Index: 14,3074 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...