Benzamidine hydrochloride hydrate, (H₂O 10-14%) 98%

Supplier: Thermo Scientific Chemicals
Warning

A16002-06 A16002-14 A16002-22
CAAAA16002-06EA 42.66 CAD
CAAAA16002-06 CAAAA16002-14 CAAAA16002-22
Benzamidine hydrochloride hydrate, (H₂O 10-14%) 98%
Benzamidine hydrochloride hydrate

A proteolytic inhibitor and apoptosis reagent


Purity: 98% CAS No.: 206752-36-5 Molecular Formula: C7H8N26.HCl.xH2O Formula Weight: 156.62(anhy) Melting Point: 82-85° Beilstein Reference: 3594959

Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR