1,3-Dimethyl-2-imidazolidinone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Dimethylimidazolidin-2-one
A16001-22
A16001-36
CAAAA16001-22EA
71.26
CAD
CAAAA16001-22
CAAAA16001-36
1,3-Dimethyl-2-imidazolidinone 98%
1,3-Dimethyl-2-imidazolidinone
Purity: 98%
CAS No.: 80-73-9
Molecular Formula: C5H10N2O
Formula Weight: 114.15
Flash Point: 93°(199°F)
Refractive Index: 1.4720
Melting Point: 8°
Boiling Point: 224-226°
Beilstein Reference: 108808
Merck Reference: 14,3249
UN No.: 2810
MDL No.: MFCD00003188
Formula:
C₅H₁₀N₂O MW: 114.15 g/mol Boiling Pt: 224…226 °C Melting Pt: 8 °C Density: 1.058 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003188 CAS Number: 80-73-9 EINECS: 201-304-8 UN: 2810 ADR: 6.1,III Merck Index: 14,3249 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...