3-Chloro-4-fluorobenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chlor-3-cyan-6-fluorbenzene

A15987-06 A15987-14
CAAAAA15987-22EA 1621.76 CAD
CAAAAA15987-22 CAAAA15987-06 CAAAA15987-14
3-Chloro-4-fluorobenzonitrile 98+%
3-Chloro-4-fluorobenzonitrile

Purity: 98+% CAS No.: 117482-84-5 Molecular Formula: C7H3ClFN Formula Weight: 155.56 Melting Point: 68-72° Beilstein Reference: 4740501 UN No.: 3439 MDL No.: MFCD00015431

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 135 °C (65 -rr)
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00015431
CAS Number: 117482-84-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR