1,3,5-Trifluorobenzene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15984-22 A15984-06 A15984-14
CAAAAA15984-22EA 1651.1 CAD
CAAAAA15984-22 CAAAA15984-06 CAAAA15984-14
1,3,5-Trifluorobenzene 98+%
1,3,5-Trifluorobenzene

Purity: 98+% CAS No.: 372-38-3 Molecular Formula: C6H3F3 Formula Weight: 132.09 Flash Point: -7°(19°F) Refractive Index: 1.4150 Melting Point: -7 to -4° Boiling Point: 75-76° Beilstein Reference: 1906457 UN No.: 2387 MDL No.: MFCD00000333

Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 75…76 °C
Melting Pt: –7 to –4 °C
Density: 1.279
Flash Pt: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS Number: 372-38-3
EINECS: 206-751-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR