2',4'-Dimethoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
A15982-14
A15982-22
CAAAA15982-14EA
111.55
CAD
CAAAA15982-14
CAAAA15982-22
2',4'-Dimethoxyacetophenone 98%
2',4'-Dimethoxyacetophenone
Purity: 98%
CAS No.: 829-20-9
Molecular Formula: C10H12O3
Formula Weight: 180.20
Flash Point:
Formula:
C₁₀H₁₂O₃ Boiling Pt: 287…289 °C Melting Pt: 39…41 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008727 CAS Number: 829-20-9 EINECS: 212-587-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...