3-Hydroxybenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger

A15981-06 A15981-14 A15981-22
CAAAA15981-06EA 57.83 CAD
CAAAA15981-06 CAAAA15981-14 CAAAA15981-22
3-Hydroxybenzyl alcohol 98%
3-Hydroxybenzyl alcohol

Purity: 98% CAS No.: 620-24-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 69-72° Boiling Point: ca 300° dec. MDL No.: MFCD00004643

Formula: C₇H₈O₂
Boiling Pt: ∼ 300 °C (dec.)
Melting Pt: 69…72 °C
Density: 1.16
Storage Temperature: Ambient
MDL Number: MFCD00004643
CAS Number: 620-24-6
EINECS: 210-633-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR