1-Octanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Octanol
A15977-0D
A15977-0F
A15977-AP
CAAAA15977-0DEA
589.34
CAD
CAAAA15977-0D
CAAAA15977-0F
CAAAA15977-AP
1-Octanol 99%
1-Octanol
Purity: 99%
CAS No.: 111-87-5
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 81°(177°F)
Physical Form: Liquid
Density: 0.825
Refractive Index: 1.4290
Appearance: Colorless
Odor: Sweet
Melting Point: -16°
Boiling Point: 193-195°
Beilstein Reference: 1697461
Merck Reference: 14,6751
MDL No.: MFCD00002988
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 193…195 °C Melting Pt: –16 °C Density: 0.825 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 14,6751 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...