Methyl cinnamate predominantly trans 99%
Supplier: Thermo Scientific Chemicals
A15975-0E
A15975-22
A15975-36
CAAAA15975-0EEA
788.42
CAD
CAAAA15975-0E
CAAAA15975-22
CAAAA15975-36
Methyl cinnamate predominantly trans 99%
Methyl cinnamate
Purity: 99%
CAS No.: 103-26-4
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point: 123°(253°F)
Physical Form: Cryst solid
Density: 1.042
Refractive Index: 1.5770
Appearance: White
Melting Point: 34-37°
Boiling Point: 262-264°
Beilstein Reference: 386468
Merck Reference: 14,2299
MDL No.: MFCD00008458
Formula:
C₁₀H₁₀O₂ Boiling Pt: 262…264 °C Melting Pt: 34…37 °C Density: 1.042 Flash Pt: 123 °C(253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008458 CAS Number: 103-26-4 EINECS: 203-093-8 Merck Index: 14,2299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...