Nicotinamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15970-22
A15970-0B
A15970-30
CAAAAA15970-22EA
43.99
CAD
CAAAAA15970-22
CAAAA15970-0B
CAAAA15970-30
Nicotinamide 99%
Nicotinamide
The amide of nicotinic acid (vitamin B-3)
Purity: 99%
CAS No.: 98-92-0
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Flash Point: 182°(360°F)
Physical Form: Powder
Density: 1.40
Appearance: White
Odor: Odorless
Melting Point: 128-132°
Beilstein Reference: 383619
Merck Reference: 14,6523
MDL No.: MFCD00006395
The amide of nicotinic acid (vitamin B-3)
Formula:
C₆H₆N₂O Boiling Pt: 334 °C (1013 hPa) Melting Pt: 128…132 °C Density: 1.40 Flash Pt: 182 °C(360 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006395 CAS Number: 98-92-0 EINECS: 202-713-4 Merck Index: 14,6523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...