1,8-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

A15966-0B A15966-18 A15966-30
CAAAA15966-0BEA 1080.72 CAD
CAAAA15966-0B CAAAA15966-18 CAAAA15966-30
1,8-Diaminonaphthalene 97%
1,8-Diaminonaphthalene

Purity: 97% CAS No.: 479-27-6 Molecular Formula: C10H10N2 Formula Weight: 158.20 Flash Point: 195°(383°F) Melting Point: 61-65° Boiling Point: 205°/12mm Beilstein Reference: 742841 Merck Reference: 14,6372 MDL No.: MFCD00004033

Formula: C₁₀H₁₀N₂
Boiling Pt: 205 °C/12 mm
Melting Pt: 61…65 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 195 °C (383 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
EINECS: 207-529-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,6372

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR