4-Amino-N,N-dimethylaniline 96%

Supplier: Thermo Scientific Chemicals
Danger

A15962-0B A15962-18 A15962-30
CAAAA15962-0BEA 1275.06 CAD
CAAAA15962-0B CAAAA15962-18 CAAAA15962-30
4-Amino-N,N-dimethylaniline 96%
4-Amino-N,N-dimethylaniline

Purity: 96% CAS No.: 99-98-9 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 130°(266°F) Melting Point: 34-38° Boiling Point: 262° Beilstein Reference: 508105 Merck Reference: 14,3254 UN No.: 2811 MDL No.: MFCD00007860

Formula: C₈H₁₂N₂
Boiling Pt: 262 °C
Melting Pt: 34…38 °C
Density: 1.09
Flash Pt: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007860
CAS Number: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,3254

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR