Trimesic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
A15947-22
A15947-36
A15947-18
A15947-30
CAAAAA15947-22EA
136.67
CAD
CAAAAA15947-22
CAAAAA15947-36
CAAAA15947-18
CAAAA15947-30
Trimesic acid 98%
Trimesic acid
Purity: 98%
CAS No.: 554-95-0
Molecular Formula: C9H6O6
Formula Weight: 210.14
Flash Point: 328°(622°F)
Melting Point: 374-376°
Beilstein Reference: 2053080
MDL No.: MFCD00002517
Formula:
C₉H₆O₆ MW: 210.14 g/mol Melting Pt: 374…376 °C Flash Pt: 328 °C (622 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002517 CAS Number: 554-95-0 EINECS: 209-077-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...