Sodium benzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid sodium salt

A15946-30 A15946-0C A15946-0E A15946-36
CAAAAA15946-30EA 39.18 CAD
CAAAAA15946-30 CAAAA15946-0C CAAAA15946-0E CAAAA15946-36
Sodium benzoate 99%
Sodium benzoate

MDL: MFCD00012463 Beilstein: 3572467 Soluble in water, alcohol


Purity: 99% CAS No.: 532-32-1 Molecular Formula: C7H5NaO2 Formula Weight: 144.11 Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 300° Solubility: Soluble in water, alcohol Beilstein Reference: 3572467 Merck Reference: 14,8582 MDL No.: MFCD00012463

Formula: C₆H₅COONa
Melting Pt: >300 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 14,8582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR