4-Bromo-α,α,α-trifluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15942-06
A15942-14
A15942-22
CAAAA15942-06EA
68.26
CAD
CAAAA15942-06
CAAAA15942-14
CAAAA15942-22
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Purity: 99%
CAS No.: 402-43-7
Molecular Formula: C7H4BrF3
Formula Weight: 225.01
Flash Point: 60°(140°F)
Refractive Index: 1.4730
Boiling Point: 154-155°
Beilstein Reference: 2045666
UN No.: 1993
MDL No.: MFCD00000398
Formula:
C₇H₄BrF₃ Boiling Pt: 154…155 °C Density: 1.630 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000398 CAS Number: 402-43-7 EINECS: 206-943-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...