trans-2,3-Dimethoxycinnamic acid predominantly trans 98+%
Supplier: Thermo Scientific Chemicals
A15936-30
A15936-09
A15936-18
CAAAAA15936-30EA
1019.1
CAD
CAAAAA15936-30
CAAAA15936-09
CAAAA15936-18
trans-2,3-Dimethoxycinnamic acid predominantly trans 98+%
trans-2,3-Dimethoxycinnamic acid
Purity: 98+%
CAS No.: 7345-82-6
Molecular Formula: C11H12O4
Formula Weight: 208.21
Melting Point: 179-183°
Beilstein Reference: 2695122
Formula:
C₁₁H₁₂O₄ |
MDL Number:
MFCD00004376 CAS Number: 7345-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...