2,6-Dibromo-4-isopropylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

A15932-22 A15932-06 A15932-14
CAAAAA15932-22EA 1004.88 CAD
CAAAAA15932-22 CAAAA15932-06 CAAAA15932-14
2,6-Dibromo-4-isopropylaniline 98%
2,6-Dibromo-4-isopropylaniline

Purity: 98% CAS No.: 10546-65-3 Molecular Formula: C9H11Br2N Formula Weight: 293.01 Melting Point: 45-47° MDL No.: MFCD00015475

Formula: C₉H₁₁Br₂N
MW: 293 g/mol
Boiling Pt: 316
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00015475
CAS Number: 10546-65-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR