4-tert-Butyl-1-bromobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15924-36
A15924-14
A15924-22
CAAAAA15924-36EA
1000.14
CAD
CAAAAA15924-36
CAAAA15924-14
CAAAA15924-22
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene
Purity: 97%
CAS No.: 3972-65-4
Molecular Formula: C10H13Br
Formula Weight: 213.12
Flash Point: 97°(206°F)
Refractive Index: 1.5330
Melting Point: 15-16°
Boiling Point: 231-232°
Beilstein Reference: 1859117
MDL No.: MFCD00000108
Formula:
C₁₀H₁₃Br Boiling Pt: 231…232 °C Melting Pt: 15…16 °C Density: 1,229 Flash Pt: 97 °C(206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000108 CAS Number: 3972-65-4 EINECS: 223-599-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...