N-Bromosuccinimide 99%

Supplier: Thermo Scientific Chemicals
Warning

A15922-0B A15922-0I A15922-30
CAAAA15922-0BEA 241.69 CAD
CAAAA15922-0B CAAAA15922-0I CAAAA15922-30
N-Bromosuccinimide 99%
N-Bromosuccinimide

Allylic bromination reagent. Cleaves peptides at tryptophyl residues


Purity: 99% CAS No.: 128-08-5 Molecular Formula: C4H4BrNO2 Formula Weight: 177.99 Odor: Pungent Melting Point: 175-180° dec. Beilstein Reference: 113916 Merck Reference: 14,1438 UN No.: 3261 MDL No.: MFCD00005510


Allylic bromination reagent. Cleaves peptides at tryptophyl residues

Formula: C₄H₄BrNO₂
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 175…180 °C (dec.)
Density: 2.098
Storage Temperature: Ambient
MDL Number: MFCD00005510
CAS Number: 128-08-5
EINECS: 204-877-2
UN: 3084
ADR: 8,II
Merck Index: 14,1438

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR