1-Bromo-4-hexylbenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Bromophenyl)hexane

A15918-06 A15918-14
CAAAA15918-03EA 66.66 CAD
CAAAA15918-03 CAAAA15918-06 CAAAA15918-14
1-Bromo-4-hexylbenzene 97%
1-Bromo-4-hexylbenzene

Purity: 97% CAS No.: 23703-22-2 Molecular Formula: C12H17Br Formula Weight: 241.17 Flash Point: 120°(248°F) Refractive Index: 1.5213 Boiling Point: 280° MDL No.: MFCD00061114

Formula: C₁₂H₁₇Br
Boiling Pt: 280 °C
Density: 1.179
Flash Pt: >120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061114
CAS Number: 23703-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR