2-Acetylpyrazine 99%
Supplier: Thermo Scientific Chemicals
A15917-03
A15917-06
CAAAA15917-03EA
81.84
CAD
CAAAA15917-03
CAAAA15917-06
2-Acetylpyrazine 99%
2-Acetylpyrazine
Purity: 99%
CAS No.: 22047-25-2
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 75-78°
Boiling Point: 78-79°/8mm
MDL No.: MFCD00006134
Formula:
C₆H₆N₂O Boiling Pt: 78…79 °C/8 mm Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006134 CAS Number: 22047-25-2 EINECS: 244-753-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...