α,α,α-Trifluoro-m-toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15910-0E
A15910-22
A15910-36
CAAAA15910-0EEA
485.06
CAD
CAAAA15910-0E
CAAAA15910-22
CAAAA15910-36
α,α,α-Trifluoro-m-toluidine 99%
α,α,α-Trifluoro-m-toluidine
Purity: 99%
CAS No.: 98-16-8
Molecular Formula: C7H6F3N
Formula Weight: 161.13
Flash Point: 85°(185°F)
Refractive Index: 1.4800
Odor: Recognizable
Melting Point: 5-6°
Boiling Point: 187-189°
Beilstein Reference: 387672
UN No.: 2948
MDL No.: MFCD00007797
Formula:
C₇H₆F₃N Boiling Pt: 187…189 °C Melting Pt: 5…6 °C Density: 1.295 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007797 CAS Number: 98-16-8 EINECS: 202-643-4 UN: 2948 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...