4-Bromobenzenesulfonyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15909-14
A15909-22
A15909-36
CAAAA15909-14EA
94.8
CAD
CAAAA15909-14
CAAAA15909-22
CAAAA15909-36
4-Bromobenzenesulfonyl chloride 98+%
4-Bromobenzenesulphonyl chloride
Purity: 98+%
CAS No.: 98-58-8
Molecular Formula: C6H4BrClO2S
Formula Weight: 255.52
Melting Point: 73-77°
Boiling Point: 152-154°/26mm
Merck Reference: 14,1407
UN No.: 3261
MDL No.: MFCD00007437
Formula:
C₆H₄BrClO₂S Boiling Pt: 152…154 °C/26 mm Melting Pt: 73…77 °C Density: 1.37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007437 CAS Number: 98-58-8 EINECS: 202-683-2 UN: 3261 ADR: 8,III Merck Index: 14,1407 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...