Pelargonaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Nonanal, Nonylaldehyde, Nonanal, Nonaldehyde
A15908-22
A15908-36
CAAAA15908-22EA
100.8
CAD
CAAAA15908-22
CAAAA15908-36
Pelargonaldehyde 97%
Pelargonaldehyde
Purity: 97%
CAS No.: 124-19-6
Molecular Formula: CH3(CH2)7CHO
Formula Weight: 142.24
Flash Point: 63°(145°F)
Refractive Index: 1.4240
Boiling Point: 79-81°/12mm
Beilstein Reference: 1236701
MDL No.: MFCD00007030
Formula:
C₉H₁₈O Boiling Pt: 79…81 °C/12 mm Melting Pt: –40 °C Density: 0.825 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007030 CAS Number: 124-19-6 EINECS: 204-688-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...