Methyl benzilate 98%

Supplier: Thermo Scientific Chemicals
Warning

A15907-18 A15907-30
CAAAA15907-0BEA 477.71 CAD
CAAAA15907-0B CAAAA15907-18 CAAAA15907-30
Methyl benzilate 98%
Methyl benzilate

Purity: 98% CAS No.: 76-89-1 Molecular Formula: C15H14O3 Formula Weight: 242.28 Melting Point: 73-76° Boiling Point: 187°/13mm Merck Reference: 14,1080 MDL No.: MFCD00004446

Formula: C₁₅H₁₄O₃
MW: 242.27
Boiling Pt: 320
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00004446
CAS Number: 76-89-1
EINECS: 200-991-1
Merck Index: 14,1080

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR