Isophthaloyl dichloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15904-22
A15904-36
CAAAA15904-22EA
57.35
CAD
CAAAA15904-22
CAAAA15904-36
Isophthaloyl dichloride 98%
Isophthaloyl dichloride
Purity: 98%
CAS No.: 99-63-8
Molecular Formula: C8H4Cl2O2
Formula Weight: 203.02
Flash Point: 180°(356°F)
Melting Point: 42-45°
Boiling Point: 275-278°
Beilstein Reference: 638342
UN No.: 3261
MDL No.: MFCD00000678
Formula:
C₈H₄Cl₂O₂ Boiling Pt: 275…278 °C Melting Pt: 42…45 °C Density: 1.388 Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000678 CAS Number: 99-63-8 EINECS: 202-774-7 UN: 2928 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...