(4-Nitrophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitrophenylacetic acid

A15901-14 A15901-22 A15901-36
CAAAA15901-14EA 49.45 CAD
CAAAA15901-14 CAAAA15901-22 CAAAA15901-36
(4-Nitrophenyl)acetic acid 98%
(4-Nitrophenyl)acetic acid

Purity: 98% CAS No.: 104-03-0 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 152-156° Beilstein Reference: 1911801 Merck Reference: 14,6622 MDL No.: MFCD00007383

Formula: (C₆H₄(NO₂))CH₂COOH
MW: 181.15 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007383
CAS Number: 104-03-0
EINECS: 203-168-5
Merck Index: 14,6622

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR