DL-Norvaline 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Nva-OH, 2-Aminopentanoic acid, (±)-Norvaline

A15900-22 A15900-36
CAAAA15900-22EA 128.93 CAD
CAAAA15900-22 CAAAA15900-36
DL-Norvaline 98%
DL-Norvaline

Purity: 98% CAS No.: 760-78-1 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 300° Beilstein Reference: 1721163 Merck Reference: 14,6716 MDL No.: MFCD00064420

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 223
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064420
CAS Number: 760-78-1
EINECS: 212-082-7
Merck Index: 14,6716

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR