Sodium diethyldithiocarbamate trihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

A15898-22 A15898-36
CAAAA15898-22EA 98.28 CAD
CAAAA15898-22 CAAAA15898-36
Sodium diethyldithiocarbamate trihydrate 98%
Sodium diethyldithiocarbamate trihydrate

Purity: 98% CAS No.: 20624-25-3 Molecular Formula: C5H10NNaS2.3H2O Formula Weight: 225.31 (171.28anhy) Melting Point: 94-102° Beilstein Reference: 3920507 Merck Reference: 14,3378 MDL No.: MFCD00150617

Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 94…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9 III
Merck Index: 14,3378

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR