2-Mercaptoethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15890-0B
A15890-0I
A15890-30
CAAAA15890-0BEA
162.74
CAD
CAAAA15890-0B
CAAAA15890-0I
CAAAA15890-30
2-Mercaptoethanol 98+%
2-Mercaptoethanol
Solubilizes proteins by reducing disulfide linkages
Purity: 98+%
CAS No.: 60-24-2
Molecular Formula: HSCH2CH2OH
Formula Weight: 78.13
Flash Point: 73°(163°F)
Refractive Index: 1.500
Odor: Unpleasant
Melting Point: -100°
Boiling Point: 156-158°
Beilstein Reference: 773648
Merck Reference: 14,5869
UN No.: 2966
MDL No.: MFCD00004890
Formula:
HSCH₂CH₂OH Boiling Pt: 156…158 °C Melting Pt: –100 °C Density: 1.114 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004890 CAS Number: 60-24-2 EINECS: 200-464-6 UN: 2966 ADR: 6.1,II Merck Index: 14,5869 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...