3-Bromo-o-xylene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2,3-dimethylbenzene

A15889-06 A15889-14 A15889-22
CAAAA15889-06EA 67.62 CAD
CAAAA15889-06 CAAAA15889-14 CAAAA15889-22
3-Bromo-o-xylene 99%
3-Bromo-o-xylene

Purity: 99% CAS No.: 576-23-8 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 80°(176°F) Refractive Index: 1.5600 Boiling Point: 212-214° Beilstein Reference: 2077312 MDL No.: MFCD00000069

Formula: C₈H₉Br
Boiling Pt: 212…214 °C
Density: 1.365
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR