1-Bromo-3-fluoro-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A15884-14 A15884-22
CAAAA15884-14EA 74.1 CAD
CAAAA15884-14 CAAAA15884-22
1-Bromo-3-fluoro-4-iodobenzene 98%
1-Bromo-3-fluoro-4-iodobenzene

Purity: 98% CAS No.: 105931-73-5 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Melting Point: 46-50° Beilstein Reference: 4740250 MDL No.: MFCD00010608

Formula: C₆H₃BrFI
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 46…50 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010608
CAS Number: 105931-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR