Thymine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil

A15879-14 A15879-22 A15879-36
CAAAA15879-14EA 85.79 CAD
CAAAA15879-14 CAAAA15879-22 CAAAA15879-36
Thymine 97%
Thymine

Purity: 97% CAS No.: 65-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 316-317° Beilstein Reference: 607626 Merck Reference: 14,9398 MDL No.: MFCD00006026

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Boiling Pt: 389 °C (1013 hPa)
Melting Pt: 316…317 °C
Density: 1.454 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006026
CAS Number: 65-71-4
EINECS: 200-616-1
Merck Index: 14,9398

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR