(Methyl)triphenylphosphonium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15878-0B
A15878-18
A15878-30
CAAAA15878-0BEA
701.52
CAD
CAAAA15878-0B
CAAAA15878-18
CAAAA15878-30
(Methyl)triphenylphosphonium bromide 98+%
(Methyl)triphenylphosphonium bromide
Purity: 98+%
CAS No.: 1779-49-3
Molecular Formula: CH3P(C6H5)3Br
Formula Weight: 357.24
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 231-234°
Beilstein Reference: 3599467
UN No.: 2811
MDL No.: MFCD00011804
Formula:
C₁₉H₁₈BrP MW: 357.23 g/mol Melting Pt: 232 Flash Pt: >240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011804 CAS Number: 1779-49-3 EINECS: 217-218-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...