3,4-Difluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15876-03 A15876-06 A15876-14
CAAAA15876-03EA 31.44 CAD
CAAAA15876-03 CAAAA15876-06 CAAAA15876-14
3,4-Difluorobenzoic acid 98%
3,4-Difluorobenzoic acid

Purity: 98% CAS No.: 455-86-7 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Odor: Odorless Melting Point: 121-124° Beilstein Reference: 2085848 MDL No.: MFCD00011672

Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 121…124 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7
EINECS: 207-249-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR