3,4-Dichlorobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15873-14
A15873-22
A15873-36
CAAAA15873-14EA
43.61
CAD
CAAAA15873-14
CAAAA15873-22
CAAAA15873-36
3,4-Dichlorobenzoic acid 99%
3,4-Dichlorobenzoic acid
Purity: 99%
CAS No.: 51-44-5
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 205-208°
Beilstein Reference: 2044777
Formula:
C₇H₄Cl₂O₂ Boiling Pt: 301 °C (1013 hPa) Melting Pt: 205…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002492 CAS Number: 51-44-5 EINECS: 200-099-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...