2,5-Dimethoxybenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15868-06
A15868-14
CAAAA15868-06EA
118.34
CAD
CAAAA15868-06
CAAAA15868-14
2,5-Dimethoxybenzonitrile 98%
2,5-Dimethoxybenzonitrile
Purity: 98%
CAS No.: 5312-97-0
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 80-84°
Beilstein Reference: 2691937
UN No.: 3439
MDL No.: MFCD00016375
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Boiling Pt: 283 °C (1013 hPa) Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016375 CAS Number: 5312-97-0 EINECS: 226-169-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...