2-Chlorobenzophenone 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15867-14
A15867-22
CAAAA15867-14EA
58.93
CAD
CAAAA15867-14
CAAAA15867-22
2-Chlorobenzophenone 99+%
2-Chlorobenzophenone
Purity: 99+%
CAS No.: 5162-03-8
Molecular Formula: C13H9ClO
Formula Weight: 216.67
Flash Point:
Formula:
C₁₃H₉ClO Boiling Pt: 330 °C Melting Pt: 44…47 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000558 CAS Number: 5162-03-8 EINECS: 225-936-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...